Learn More
Sigma Aldrich Fine Chemicals Biosciences Human DNA for Whole Genome
Catalog No. 502909053
Encompass
This Reference Material (RM) is intended for validation optimization and process evaluation purposes. It consists of a male whole human genome sample of Eastern European Ashkenazi Jewish ancestry from the Personal Genome Project (ID huAA53E0) and it can be used to assess performance of variant calling from genome sequencing. A unit of RM 8391 consists of a vial containing human genomic DNA extracted from a single large growth of human lymphoblastoid cell line GM24385 (labeled as HG-002) from the Coriell Institute for Medical Research (Camden NJ). SRM 8391SDS
Catalog No. 50-290-9053 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST8391
This Reference Material (RM) is intended for validation optimization and process evaluation purposes. It consists of a male whole human genome sample of Eastern European Ashkenazi Jewish ancestry from the Personal Genome Project (ID huAA53E0) and it can be used to assess performance of variant calling from genome sequencing. A unit of RM 8391 consists of a vial containing human genomic DNA extracted from a single large growth of human lymphoblastoid cell line GM24385 (labeled as HG-002) from the Coriell Institute for Medical Research (Camden NJ). SRM 8391SDS