Learn More
Biomatik Corporation Human DNA Methyltransferase 3 Like Protein (DNMT3L) ELISA Kit
Supplier:  Biomatik Corporation EKU11818
Encompass_Preferred
Human DNA Methyltransferase 3 Like Protein (DNMT3L) ELISA Kit for detecting DNMT3L in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UJW3. Gene ID: 29947. Target Synonyms: Cytosine-5-Methyltransferase 3-Like; DNA (cytosine-5)-methyltransferase 3-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5993