Learn More
Biomatik Corporation Human DNA Methyltransferase 3B (DNMT3B) ELISA Kit
Supplier:  Biomatik Corporation EKU03771
Encompass_Preferred
Human DNA Methyltransferase 3B (DNMT3B) ELISA Kit for detecting DNMT3B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBC3. Gene ID: 1789. Target Synonyms: ICF; M.HsaIIIB; DNA methyltransferase HsaIIIB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9008