Learn More
Biomatik Corporation Human DnaJ/HSP40 Homolog Subfamily C, Member 12 (DNAJC12) ELISA Kit
Supplier:  Biomatik Corporation EKU03775
Encompass_Preferred
Human DnaJ/HSP40 Homolog Subfamily C, Member 12 (DNAJC12) ELISA Kit for detecting DNAJC12 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UKB3. Gene ID: 56521. Target Synonyms: JDP1; J Domain Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4794