Learn More
Abcam Human DNase I Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502660527
Encompass_Preferred
Human DNase I Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human DNase I.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-0527 Supplier Abcam Supplier No. AB30956410X96TESTS
Human DNase I Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human DNase I.
The product is subject to the following: Abcam Restricted Use Statement