Learn More
ACROBiosystems Human IgG1 Kappa Isotype Control (mAb), carrier-free, 100 μg (ACROBiosystems)
Catalog No. NC2093166
Encompass_Preferred
ACROBiosystems Human IgG1 Kappa Isotype Control is a recombinant chimeric monoclonal antibody expressed in HEK293 cells, provided carrier-free and MALS-verified. Supplied as a 100 μg lyophilized product, it is intended as a DNP-reactive isotype control for assay development and validation in ELISA, western blot, immunoprecipitation, and related immunoassays.
  • Recombinantly expressed in HEK293 cells.
  • Carrier-free, lyophilized from PBS with trehalose as protectant.
  • Purity greater than 95% as determined by SDS-PAGE.
  • SEC-MALS verified molecular weight approximately 135-160 kDa.
  • QC-tested ELISA linear range: 1-10 ng/mL.
  • Store lyophilized at -20°C or lower; reconstituted product stable at -70°C for up to 3 months.
This item is not returnable. View return policy
Catalog No. NC2093166 Supplier ACROBiosystems Supplier No. DNPM2100UG
ACROBiosystems Human IgG1 Kappa Isotype Control is a recombinant chimeric monoclonal antibody expressed in HEK293 cells, provided carrier-free and MALS-verified. Supplied as a 100 μg lyophilized product, it is intended as a DNP-reactive isotype control for assay development and validation in ELISA, western blot, immunoprecipitation, and related immunoassays.
  • Recombinantly expressed in HEK293 cells.
  • Carrier-free, lyophilized from PBS with trehalose as protectant.
  • Purity greater than 95% as determined by SDS-PAGE.
  • SEC-MALS verified molecular weight approximately 135-160 kDa.
  • QC-tested ELISA linear range: 1-10 ng/mL.
  • Store lyophilized at -20°C or lower; reconstituted product stable at -70°C for up to 3 months.