Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DOG-1 Synthetic Peptide

Catalog No. PEP0939
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0939 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0939 Supplier Invitrogen™ Supplier No. PEP0939
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 17 amino acids near the amino terminus of human TMEM16A. PEP-0939 can be used as a blocking peptide with polyclonal antibody PA5-20825.

DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55107
Species Human
Purification Method purified
Accession Number Q5XXA6
Common Name DOG-1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2
Gene Symbol ANO1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.