Learn More
Biomatik Corporation Human Dolichyl Diphosphooligosaccharide Protein Glycosyltransferase (DDOST) ELISA Kit
Supplier:  Biomatik Corporation EKU09263
Encompass_Preferred
Human Dolichyl Diphosphooligosaccharide Protein Glycosyltransferase (DDOST) ELISA Kit for detecting DDOST in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39656. Gene ID: 1650. Target Synonyms: AGE-R1; OK/SW-cl.45; OST; OST48; WBP1; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; Oligosaccharyl transferase 48 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6185