Learn More
Biomatik Corporation Human Dopa Decarboxylase (DDC) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10922
Encompass_Preferred
Human Dopa Decarboxylase (DDC) High-sensitive ELISA Kit for detecting DDC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20711. Gene ID: 1644. Target Synonyms: AADC; Aromatic L-Amino Acid Decarboxylase; DOPA decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6584