Learn More
Biomatik Corporation Human Dopamine Receptor D1 (DRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU03781
Encompass_Preferred
Human Dopamine Receptor D1 (DRD1) ELISA Kit for detecting DRD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21728. Gene ID: 1812. Target Synonyms: DR-D1; D1R; DADR; DRD1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5880