Learn More
Biomatik Corporation Human Dopamine Transporter (DAT) ELISA Kit
Supplier:  Biomatik Corporation EKU03787
Encompass_Preferred
Human Dopamine Transporter (DAT) ELISA Kit for detecting DAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01959. Gene ID: 6531. Target Synonyms: SLC6A3; DAT1; Sodium-dependent dopamine transporter; Dopamine Active Transporter; SLC6-A3; Solute Carrier Family 6 (neurotransmitter Transporter,Dopamine) ,Member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9005