Learn More
Biomatik Corporation Human Double C2 Like Domains Beta (DOC2b) ELISA Kit
Supplier:  Biomatik Corporation EKU12050
Encompass_Preferred
Human Double C2 Like Domains Beta (DOC2b) ELISA Kit for detecting DOC2b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14184. Gene ID: 8447. Target Synonyms: DOC2BL; Double C2-like domain-containing protein beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8046