Learn More
Sino Biological Anti-DPEP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13543RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human dehydropeptidase-I / DPEP1 (rh DPEP1; Catalog#13543-H08H; P16444; Met1-Ser385). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4220