Learn More
Sino Biological Anti-DPEP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13806RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MBD2 / DPEP2 (rh MBD2 / DPEP2; Catalog#13806-H08H; AAH24021.1; Met1-Ser376). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4185