Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DPP4 (aa 425-510) Control Fragment Recombinant Protein

Catalog No. RP109167
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109167 Supplier Invitrogen™ Supplier No. RP109167
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27487
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 1803
Name Human DPP4 (aa 425-510) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10
Common Name DPP4
Gene Symbol DPP4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPGGRNLYKIQLSDYTKVTCLSCELNPERCQYYSVSFSKEAKYYQLRCSGPGLPLYTLHSSVNDKGLRVLEDNSALDKMLQNVQMP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.