Learn More
Sino Biological Recombinant Human DPP-4/CD26/DPPIV Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10688H08H50
Encompass_Preferred
A DNA sequence encoding the human DPP4 (NP_001926.2) (Asp34-Pro766) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6739