Learn More
Sino Biological Recombinant Human DPP-4/CD26/DPPIV Protein, HPLC-verified
Supplier:  Sino Biological 10688HNCH50
Encompass_Preferred
The native mature form of human DPPIV (NP_001926.2) extracellular domain (Asn 29-Pro 766) was expressed and purified.
Catalog No. 50-224-6795