Learn More
Sino Biological Anti-DPP-4/CD26/DPPIV Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10688MM05A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DPP4 / CD26 (rh DPP4 / CD26; Catalog#10688-HNCH; NP_001926.2; Asn29-Pro766) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1525