Learn More
Sino Biological Anti-DPP2 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10748MM03H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DPPII / DPP7 (rhDPPII; Catalog#10748-H08H; NP_037511.2; Met 1-Leu 492). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8746