Learn More
Sino Biological Anti-DPP2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10748MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DPP7 / DPPII / DPP2 (rh DPP7 / DPPII / DPP2; Catalog#10748-H08H; NP_037511.2; Met1-Leu492). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8769