Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DPPA4 (aa 6-97) Control Fragment Recombinant Protein

Catalog No. RP96328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96328 Supplier Invitrogen™ Supplier No. RP96328
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (39%), Rat (39%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83266. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The DPPA4 gene encodes developmental pluripotency associated 4, a protein that plays a crucial role in maintaining stem cell pluripotency and facilitating early embryonic development. DPPA4 is primarily expressed in pluripotent stem cells, such as embryonic stem cells, where it contributes to the regulation of gene expression and chromatin structure necessary for pluripotency and differentiation. This protein is also involved in the establishment of epigenetic marks that influence developmental pathways, ensuring the proper formation of tissue types during embryogenesis. DPPA4 functions in stem cell renewal and differentiation, impacting cellular processes that are fundamental to early development. Beyond its roles in normal physiology, DPPA4 has been studied in the context of oncology, where its expression is often upregulated in certain cancers, indicating its potential as a biomarker for tumorigenic stem cell-like properties. Research continues to explore DPPA4's mechanisms and interactions to understand its contributions to development and its implications in disease states, particularly in cancer biology and regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L190
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 55211
Name Human DPPA4 (aa 6-97) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2410091M23Rik; C76608; developmental pluripotency associated 4; developmental pluripotency-associated protein 4; Dppa4; ECAT15-1; Embryonic stem cell-associated transcript 15-1 protein; LOC685378; similar to developmental pluripotency associated 4 isoform 1
Common Name DPPA4
Gene Symbol DPPA4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ASSTSMEKAKGKEWTSTEKSREEDQQASNQPNSIALPGTSAKRTKEKMSIKGSKVLCPKKKAEHTDNPRPQKKIPIPPLPSKLPPVNLIHRD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.