Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DPPII/QPP/DPP7 ELISA Kit

Catalog No. EH160RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH160RBX5 5 x 96 Tests
EH160RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH160RBX5 Supplier Invitrogen™ Supplier No. EH160RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human DPPII/QPP/DPP7 quantitates human DPPII/QPP/DPP7 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DPPII/QPP/DPP7.

The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHL4
Assay Range 0.11-25 ng/mL
Assay Sensitivity 0.11 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidasecarboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase;
Gene ID (Entrez) 29952
Gene Symbol DPP7
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target DPP2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.