Learn More
ACROBiosystems ACROBiosystems Human DR3 / TNFRSF25 Protein, His Tag
Supplier:  ACROBiosystems TN5H52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human DR3 / TNFRSF25, His Tag (TN5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 199 (Accession # Q93038-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 30-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7378