Learn More
Description
This peptide corresponds to 20 amino acids near the carboxy terminus of human DR5. PEP-0021 can be used as a blocking peptide with polyclonal antibody PA5-19895.
Specifications
Specifications
| Gene ID (Entrez) | 8795 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O14763 |
| Common Name | TRAIL-R2 (DR5) |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.