Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DR6 Synthetic Peptide

Catalog No. PEP0055 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0055 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0055 Supplier Invitrogen™ Supplier No. PEP0055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 15 amino acids of human DR6. PEP-0055 can be used as a blocking peptide with polyclonal antibody PA5-19930.

DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kappa-B and JNK.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 27242
Species Human
Purification Method purified
Accession Number O75509
Common Name DR6
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias AA959878; BM-018; CD358; Death receptor 6; Death receptor-6; DR6; MGC31965; R74815; sCD358; sDR6; soluble CD358; soluble DR6; TNF receptor superfamily member 21; TNFR-related death receptor 6; TNFR-related death receptor-6; Tnfrsf21; TR7; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21; UNQ437/PRO868
Gene Symbol TNFRSF21
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.