Learn More
ACROBiosystems ACROBiosystems Human DR6 / TNFRSF21 Protein, Fc Tag
Catalog No. 502106769
Encompass_Preferred
Recombinant Protein;200UG;Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession # AAH17730).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 80-95kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6769 Supplier ACROBiosystems Supplier No. TN1H5252200UG
Recombinant Protein;200UG;Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession # AAH17730).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 80-95kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.