Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Draxin/Neucrin ELISA Kit

Catalog No. EH161RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH161RBX5 5 x 96 Tests
EH161RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH161RBX5 Supplier Invitrogen™ Supplier No. EH161RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Draxin/Neucrin quantitates human Draxin/Neucrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Draxin/Neucrin.

Chemorepulsive axon guidance protein required for the development of spinal cord and forebrain commissures. Acts as a chemorepulsive guidance protein for commissural axons during development. Able to inhibit or repel neurite outgrowth from dorsal spinal cord. Inhibits the stabilization of cytosolic beta-catenin (CTNNB1) via its interaction with LRP6, thereby acting as an antagonist of Wnt signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NBI3
Assay Range 28-7000 pg/mL
Assay Sensitivity 28 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2610109H07Rik; AGPA3119; C1orf187; Dorsal inhibitory axon guidance protein; dorsal repulsive axon guidance protein; Draxin; FLJ34999; MGC117222; Neucrin; neural tissue-specific cysteine-rich protein; PSEC0258; UNQ3119; UNQ3119/PRO102682610109H07Rik; AGPA3119; C1orf187; Dorsal inhibitory axon guidance protein; dorsal repulsive axon guidance protein; Draxin; FLJ34999; MGC117222; Neucrin; neural tissue-specific cysteine-rich protein; PSEC0258; UNQ3119; UNQ3119/PRO10268;
Gene ID (Entrez) 374946
Gene Symbol DRAXIN
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Draxin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.