Learn More
Raybiotech Inc Human Draxin/Neucrin ELISA Kit
Supplier:  Raybiotech Inc ELHDRAXIN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Draxin.
Catalog No. 50-194-9199