Learn More
Raybiotech Inc Quantibody Human Dry Eye Disease Array 1 Kit.
Supplier:  Raybiotech Inc QAHDED12
Encompass_Preferred
Detects 40 DED associated cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: EGF, EGFR, Eotaxin-1 (CCL11), GCSF, GRO alpha/beta/gamma, HB-EGF, ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-16, IL-17A, IL-2, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-8 (CXCL8), IP-10 (CXCL10), Lipocalin-2 (NGAL), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), MMP-9, RANTES (CCL5), TGF beta 1, TIMP-1, TIMP-2, TNF alpha, VEGF-A
Catalog No. 50-195-0830