Learn More
Raybiotech Inc Human Desmocollin-3 ELISA Kit
Supplier:  Raybiotech Inc ELHDSC31
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.16 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Desmocollin-3. Other species not yet determined.
Catalog No. 50-194-9201