Learn More
Sino Biological Anti-Desmocollin 2/DSC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10809RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DSC2 (rh DSC2; Catalog#10809-H08H; Q02487-1; Met 1-Arg 684). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8908