Learn More
Sino Biological Recombinant Human Desmoglein-2 Protein (His Tag), HPLC-verified
Catalog No. 502993560
Encompass_Preferred
A DNA sequence encoding the Human DSG2 (Q14126) (Ala50-Gly609) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3560 Supplier Sino Biological Supplier No. 13684H08H100
A DNA sequence encoding the Human DSG2 (Q14126) (Ala50-Gly609) was expressed with a polyhistidine tag at the C-terminus.