Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DUSP11 (aa 48-197) Control Fragment Recombinant Protein

Catalog No. RP105747
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105747 Supplier Invitrogen™ Supplier No. RP105747
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64985. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DUSP11 (dual specificity phosphatase 11 (RNA/RNP complex 1-interacting)) is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product is localized to the nucleus, and is novel in that it binds directly to RNA and splicing factors, and thus suggested to participate in nuclear mRNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75319
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 8446
Name Human DUSP11 (aa 48-197) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2010300F21Rik; AI481497; dual specificity phosphatase 11; dual specificity phosphatase 11 (RNA/RNP complex 1-interacting); dual specificity protein phosphatase 11; DUSP11; Phosphatase that interacts with RNA/RNP complex 1; PIR1; RNA/RNP complex 1-interacting; RNA/RNP complex-1-interacting phosphatase; RNA/RNP complex-1-interacting phosphatase-like protein; RNA/RNP complex-interacting phosphatase; serine/threonine specific protein phosphatase
Common Name DUSP11
Gene Symbol DUSP11
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MSQWHHPRSGWGRRRDFSGRSSAKKKGGNHIPERWKDYLPVGQRMPGTRFIAFKVPLQKSFEKKLAPEECFSPLDLFNKIREQNEELGLIIDLTYTQRYYKPEDLPETVPYLKIFTVGHQVPDDETIFKFKHAVNGFLKENKDNDKLIGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.