Learn More
Biomatik Corporation Human Dynamin 1 Like Protein (DNM1L) ELISA Kit
Supplier:  Biomatik Corporation EKU12081
Encompass_Preferred
Human Dynamin 1 Like Protein (DNM1L) ELISA Kit for detecting DNM1L in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00429. Gene ID: 10059. Target Synonyms: DRP1; DLP1; DVLP; DYMPLE; HDYNIV; VPS1; Dynamin-related protein 1; Dnm1p/Vps1p-like protein; Dynamin family member proline-rich carboxyl-terminal domain less. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8037