Learn More
Biomatik Corporation Human Dynamin 2 (DNM2) ELISA Kit
Supplier:  Biomatik Corporation EKU03808
Encompass_Preferred
Human Dynamin 2 (DNM2) ELISA Kit for detecting DNM2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50570. Gene ID: 1785. Target Synonyms: CMTDI1; CMTDIB; DYN2; DYNII; Cytoskeletal Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6847