Learn More
Biomatik Corporation Human Dynein, Axonemal, Heavy Chain 11 (DNAH11) ELISA Kit
Supplier:  Biomatik Corporation EKU03810
Encompass_Preferred
Human Dynein, Axonemal, Heavy Chain 11 (DNAH11) ELISA Kit for detecting DNAH11 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96DT5. Gene ID: 8701. Target Synonyms: DNAHBL; DNAHC11; DNHBL; DPL11; Dynein,Heavy Chain Beta-Like; Axonemal beta dynein heavy chain 11; Ciliary dynein heavy chain 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6392