Learn More
Biomatik Corporation Human Dynein, Cytoplasmic 1, Heavy Chain 1 (DYNC1H1) ELISA Kit
Supplier:  Biomatik Corporation EKU03811
Encompass_Preferred
Human Dynein, Cytoplasmic 1, Heavy Chain 1 (DYNC1H1) ELISA Kit for detecting DYNC1H1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14204. Gene ID: 1778. Target Synonyms: DHC1; DHC1a; DNCH1; DNCL; DNECL; DYHC; Dnchc1; HL-3; p22; Dynein heavy chain, cytosolic; Cytoplasmic dynein heavy chain 1; Cytoplasmic dynein 1 heavy chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6445