Learn More
Biomatik Corporation Human Dystrobrevin Beta (DTNb) ELISA Kit
Supplier:  Biomatik Corporation EKU03813
Encompass_Preferred
Human Dystrobrevin Beta (DTNb) ELISA Kit for detecting DTNb in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60941. Gene ID: 1838. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6848