Learn More
BIOHIPPO Human E-Cad Microsample
SDP
Catalog No. 5000314910
Encompass
Human E-Cad Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of E-Cad in serum plasma urine tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human E-Cad Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human E-Cad Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human E-Cad biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14910 Supplier BIOHIPPO Supplier No. BHE15212724
Human E-Cad Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of E-Cad in serum plasma urine tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human E-Cad Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human E-Cad Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human E-Cad biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color