Learn More
Sino Biological Human E-Cadherin Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785277
Encompass_Preferred
The Human E-Cadherin / CDH1 / E-cad / CD324 ELISA Pair Set is for the quantitative determination of Human E-Cadherin / CDH1 / E-cad / CD324.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human E-Cadherin / CDH1 / E-cad / CD324 coated on a 96-well plate. Standards and samples are added to the wells, and any Human E-Cadherin / CDH1 / E-cad / CD324 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human E-Cadherin / CDH1 / E-cad / CD324 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human E-Cadherin / CDH1 / E-cad / CD324 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85277 Supplier Sino Biological Supplier No. SEK102045
The Human E-Cadherin / CDH1 / E-cad / CD324 ELISA Pair Set is for the quantitative determination of Human E-Cadherin / CDH1 / E-cad / CD324.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human E-Cadherin / CDH1 / E-cad / CD324 coated on a 96-well plate. Standards and samples are added to the wells, and any Human E-Cadherin / CDH1 / E-cad / CD324 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human E-Cadherin / CDH1 / E-cad / CD324 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human E-Cadherin / CDH1 / E-cad / CD324 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.