Learn More
ACROBiosystems ACROBiosystems Human E-Selectin / CD62E Protein, Fc Tag
Catalog No. 502106584
Encompass_Preferred
Recombinant Protein;100UG;Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Phe 556 (Accession # NP_000441.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.8 kDa. The protein migrates as 96-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6584 Supplier ACROBiosystems Supplier No. CDEH5250100UG
Recombinant Protein;100UG;Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Phe 556 (Accession # NP_000441.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.8 kDa. The protein migrates as 96-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.