Learn More
Raybiotech Inc Human E-Selectin ELISA Kit
Supplier:  Raybiotech Inc ELHESELECTIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 30 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: Angiopoietin-1, Angiostatin , B7-1, BMP-7, CD14, CD30, CD40, CD40 Ligand, CTLA-4, CXCL16, Dkk-4 , DR6, Endostatin, Follistatin, HB-EGF, HVEM, ICAM-2, IGF-II, IL-10 Ra, IL-10 Rb, IL-18, IL-9 , IL-2 Ra, IL-2 Rb, IL-5 Ra, LAP, L-Selectin, M-CSF R, MMP-1, 2, 3, 7, 8, 9, 10 and 12, PDGF-AB, SDF-1b, Tie-1, Tie-2, TIMP-3.
Catalog No. 50-194-9234