Learn More
Sino Biological Anti-EBAG9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310899T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human EBAG9 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1634