Learn More
Sino Biological Anti-ECD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200315T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human ECD fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3968