Learn More
Biomatik Corporation Human Echinoderm Microtubule Associated Protein Like Protein 2 (EML2) ELISA Kit
Supplier:  Biomatik Corporation EKU03827
Encompass_Preferred
Human Echinoderm Microtubule Associated Protein Like Protein 2 (EML2) ELISA Kit for detecting EML2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95834. Gene ID: 24139. Target Synonyms: EMAP2; ELP70; Echinoderm MT-Associated Protein (EMAP)-Like Protein 70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6849