Learn More
Sino Biological Anti-ECM1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10362MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ECM1 (rh ECM1 ; Catalog#10362-H08H; NP_004416.1; Met1-Glu540). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2017