Learn More
Sino Biological Recombinant Anti-ECM1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10362R03020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ECM1 (rh ECM1 ; Catalog#10362-H08H; NP_004416.1; Met1-Glu540).
Catalog No. 50-240-7740