Learn More
BIOHIPPO Human ECRG4 Microsample
SDP
Catalog No. 5000314979
Encompass
Human ECRG4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECRG4 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ECRG4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ECRG4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ECRG4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14979 Supplier BIOHIPPO Supplier No. BHE15211044
Human ECRG4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECRG4 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ECRG4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ECRG4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ECRG4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color