Learn More
Sino Biological Anti-ECSIT Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14497RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ECSIT (rh ECSIT; Catalog#14497-H07E; Q9BQ95-1; Glu246-Ser431). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1214