Learn More
Biomatik Corporation Human Ectodysplasin A2 Receptor (EDA2R) ELISA Kit
Supplier:  Biomatik Corporation EKU08482
Encompass_Preferred
Human Ectodysplasin A2 Receptor (EDA2R) ELISA Kit for detecting EDA2R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HAV5. Gene ID: 60401. Target Synonyms: XEDAR; EDA-A2R; EDAA2R; TNFRSF27; X-linked ectodysplasin-A2 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5787